2,3,5-Triphenyltetrazolium chloride (TTC) solution 1%, not sterile

Supplier: BD

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

231121
90002-202EA 103.57 USD
90002-202
2,3,5-Triphenyltetrazolium chloride (TTC) solution 1%, not sterile
2,3,5-Triphenyltetrazolium chloride

Appearance: Colorless to slight amber, clear solution

Formula: C₁₉H₁₅ClN₄
MDL Number: MFCD00011963
CAS Number: 298-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR