BD Falcon™ SWUBE™ Collection and Transport System, BD Diagnostics
Supplier: BD
220690
90001-034CS 397.29 USD
90001-034 90001-128 90004-028 90001-094 90001-126
BD Falcon™ SWUBE™ Collection and Transport System, BD Diagnostics
Swabs Swabs for Sampling
Combination swabs and tubes provide a simple and effective method of collection, transporting, protecting, and storing specimens
Sterile, individually wrapped tubes are provided without media, but can be used with a wide array of reagents, media, or solutions. The 14.6–15cm (53/4–6”) long shafts reach touch sampling sites. Available with screw caps or friction caps