Sampling Swab for Environmental Monitoring, Sterile Pack, BD Diagnostics
Supplier: BD
220518
90001-119PK 1823.62 USD
90001-119
Sampling Swab for Environmental Monitoring, Sterile Pack, BD Diagnostics
Swabs Swabs for Sampling
Each sterile pack sampling swab unit is ready-to-use and comprised of a peel open pouch containing an environmental sampling swab in a tube with approximately 10 mL rinse solution
  • Ready-to-use sterile swab in a pre-filled tube of rinse solution
  • Use for surface and equipment sampling
  • Double-wrapped and gamma irradiated
The pre-filled rinse solution is a general-purpose isotonic solution with neutralizing agents for the maintenance of microorganisms and the neutralization of disinfectants. The double pouch design is gamma-irradiated.
Designed to sample a variety of surface textures and equipment with the advantage of sampling hard-to-reach areas such as equipment crevices and inside pipe work.