RODAC™ Plate, Specialty Petri Dish, Sterile, BD Biosciences
Supplier: BD
210340
90000-004CS 706.12 USD
90000-004
RODAC™ Plate, Specialty Petri Dish, Sterile, BD Biosciences
Culture Dishes Petri Dishes
RODAC® (Replicate Organism Detection and Counting) was developed for environmental studies to determine levels of microbial contamination of surfaces
When plate is filled with 12–13mL of media, the media surface is raised to a convex shape. Area of agar bed is approximately 25cm2 with Quebec-style grids of one centimeter squares. A cake-box type lid protects the agar. Polystyrene, with cover. Packaged 20 per sleeve.