Amplitude™ Kappa™ Wipes, Contec®
Supplier: Contec
AMLP0003
89800-688CS 842.6 USD
89800-688 89800-690
Amplitude™ Kappa™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
A special hydroentangleing process washes the Amplitude™ Kappa™ fabric, made of 58% polyester and 42% lyocell (high quality rayon), to provide higher levels of cleanliness, sorbency, and softness compared to similar nonwoven wipes.
  • Recommended for ISO Class 5 environments
  • Hydroentangled process provides a soft texture for sensitive applications
  • Higher levels of cleanliness compared to similar nonwoven wipes
  • Highly sorbent with both water and solvents
  • Low levels of particles and fibres
  • Ideal for wiping delicate surfaces and sensitive components
Featuring a fine aperture and low levels of fibers and particles, these wipes are ideal for wiping delicate surfaces and sensitive components, as well as for other critical applications.
Amplitude™ Kappa™ wipes are highly sorbent with both water and solvents and have low levels of particles and fibres. Ideal for wiping delicate surfaces such as optics, plastic, and sensitive components, or for polishing metal surfaces.