Multi-Element ICP and ICP-MS Certified Reference Standards, ARISTAR®, VWR Chemicals BDH®
BDH® ARISTAR®
89800-582 BDHCMS-2 89800-664 89800-594 89800-562 BDHCMS-4 BDHCMS-5 89800-672 89800-654 89800-600 89800-668 BDHCMS-3 89800-666 89800-592 89800-568 89800-558 BDHCCS-5 89800-574 BDHIVICPM71B BDHCCS-2 89800-598 89800-578 BDHCCS-2 89800-610 89800-624 BDHIVICPM71C 89800-626 89800-648 BDHCMS-1 89800-576 89800-560 89800-584 89800-548 BDHCMS-4 BDHCMS-3 89800-646 89800-606 89800-556 89800-554 89800-632 89800-670 89800-620 BDHIVICPM71D 89800-566 89800-634 89800-662 89800-636 89800-630 BDHCCS-5 BDHCCS-1 89800-596 89800-644 BDHCCS-4 BDHCCS-4 89800-622 BDHCCS-6 89800-658 89800-602 89800-570 89800-650 89800-564 BDHIVICPM71C BDHIVICPM71A 89800-608 BDHCMS-2 89800-660 BDHIVICPM71A BDHIVICPM71B 89800-552 89800-640 89800-656 BDHCCS-6 BDHIVICPM71D 89800-550 BDHCCS-1 89800-628 89800-612 BDHCMS-1 89800-580 89800-638 BDHCMS-5 89800-652 89800-614 89800-604 89800-572 89800-586 89800-588 89800-616 89800-642 89800-618 89800-590
89800-572EA 130.63 USD
89800-572 89800-650 82026-112 89800-570 82026-110 89800-576 89800-654 89800-610 89800-574 89800-652 89800-608 89800-646 89800-602 82026-108 82026-106 89800-600 89800-644 89800-568 82026-104 89800-606 CA82026-114 82026-102 CA82026-112 89800-648 89800-604 82026-100 89800-660 CA82026-110 89800-584 89800-582 89800-664 89800-620 89800-588 89800-662 89800-586 89800-580 82025-930 82025-932 82025-934 CA82026-108 89800-658 CA82026-106 89800-614 89800-578 CA82026-104 89800-656 89800-612 CA82026-102 89800-618 CA82026-100 89800-616 89800-594 89800-550 89800-672 89800-592 82026-098 89800-670 89800-598 89800-554 89800-632 89800-596 89800-552 89800-630 89800-590 CA82025-930 CA82025-932 89800-668 89800-624 89800-548 82025-926 CA82025-934 89800-666 89800-622 82025-928 89800-628 89800-626 89800-562 CA82026-098 89800-560 89800-642 89800-566 89800-640 89800-564 89800-558 89800-636 89800-556 CA82025-926 89800-634 CA82025-928 82026-114 89800-638
Multi-Element ICP and ICP-MS Certified Reference Standards, ARISTAR®, VWR Chemicals BDH®
Stock multi-element ICP and ICP-MS blends can be found grouped for stability and ease of use as well as commonly used Certified Reference Materials (CRMs) recommended by many instrument manufactures
For the pharmaceutical industry, the range includes a line of CRMs that comply with the general chapters on elemental impurity USP <232> limits and USP<233> procedures. These multi-element CRMs are available for precious metals, oral elemental and parenteral element impurities.
These ICP and ICP-MS Certified Reference Materials (CRMs) are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs.
Each multi-element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS) and one year expiration. Specific lot C of A’s can also be found online. Most products are available in stock at the 125mL and 500mL sizes packaged in plastic LDPE bottles.
Ordering information: Uranium is depleted. The isotopic abundance is on the certificate of analysis.