Cobra® Single Channel Pipettors, Biotix
Supplier: Avantor
89525-046EA 492.55 USD
89525-046 89525-052 10770-390 10770-328 10770-324 10770-322 10813-588 10813-592
Cobra® Single Channel Pipettors, Biotix
Pipettes Micropipettes
The Cobra® Pipettor system offers an ergonomic solution to single channel variable volume pipetting applications.
Constructed for accuracy, the Cobra® Pipettor system is an innovative solution, and when paired with Biotix tips provides unmatched accuracy and precision. Additional features include easy on-site calibration (calibration key included), a controlled volume setting that works to prevent accidental volume changes, and a stainless steel piston to enable better accuracy and precision over time for long-lasting results. These pipettors work with most tips on the market (tested for perfect fit with Biotix manual tips). All units are fully autoclavable (120 °C/0.1 MPa/20 min.).
These devices feature a soft spring system for extremely low pipetting forces and an adjustable tip-ejector button to provide the best fit and ejection for right- or left-handed use in any hand size. A large, arched finger hook design provides all-day comfort and reduces tension for all hand sizes. The higher hook positions the thumb much closer to the push button, further reducing overall pipetting forces. All Cobra® Single Channel Pipettors are lightweight with durable construction.
Ordering information: The Cobra® Pipettor is supplied with a one-year manufacturer's warranty.