VWR® Disposable 3-Ply Face Mask
Supplier: VWR International
89523-476
89523-476CS 65.2 USD
89523-476
VWR® Disposable 3-Ply Face Mask
Masks Hygiene Masks
Disposable 3-ply face masks are equipped with earloops for a secure and comfortable fit.
Face masks provide a Bacterial Filtration Efficiency of >98%. These yellow masks are packaged 70 per pack.