VWR® Sterile Hood, 10⁻⁶ SAL
Supplier: VWR International
89523-474
89523-474CS 548.98 USD
89523-474
VWR® Sterile Hood, 10⁻⁶ SAL
Disposable Garments
The Sterile hood is manufactured from a specially formulated breathable microporous fabric that provides significantly fluid and barrier protection
  • Excellent breathability and water vapor transmission rate
  • Lowest level of particle shedding
  • Durable yet soft, cloth-like fabric
  • Gamma irradiated to SAL of 10-6
This hood is rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Hoods offer a BFE of over 98%. Hoods measure 17.8cm (7") at the head position. 60g SMS fabric.
It exhibits excellent water vapor transmission to optimize user comfort. The hood is gamma irradiated to a Sterility Assurance Level (SAL) of 10-6 to address the concern of bio-burden contamination.
Certifications: Manufactured in an ISO Class 8 (FED-STD-209E Class 100,000/M3.5) cleanroom. 
Ordering information: Packed individually in sealed polyethylene bags contained within a larger carton liner. Each case is supplied with a certificate of radiation.