VWR® Advanced Disposable Hood
Supplier: VWR International
89523-428
89523-428CS 163.31 USD
89523-428
VWR® Advanced Disposable Hood
Disposable Garments
Disposable non-woven hood is made of 43gsm SMS fabric
  • Nonwoven, 43gsm SMS fabric
  • Reliable protection in laboratory, electrical, pharmaceutical, food, industrial, and cleanroom applications
  • 27.9Wx41.9Lcm (11x16.5")
Disposable non-woven hood is made of 43gsm SMS fabric. Full garment dimensions are 27.9W x 41.9L cm (11 x 16.5").
Hoods provide reliable protection in laboratory, electrical, pharmaceutical, food, industrial, and cleanroom applications. Full garment dimensions are 27.9W x 41.9L cm (11 x 16.5").
Certifications: ISO9001