Anti-TFAP2B Rabbit Polyclonal Antibody

Supplier: Abgent
AP13256B
89521-718EA 646.4 USD
89521-718
Anti-TFAP2B Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: 1:1000

Type: Primary
Antigen: TFAP2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR