Anti-ATG4C Mouse Monoclonal Antibody [clone: 287CT12.2.2]

Supplier: Abgent
AM1933B
89521-612EA 646.72 USD
89521-612
Anti-ATG4C Mouse Monoclonal Antibody [clone: 287CT12.2.2]
Antibodies
Western Blot: 1:100,

Type: Primary
Antigen: ATG4C
Clonality: Monoclonal
Clone: 287CT12.2.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR