Anti-DICER1 Mouse Monoclonal Antibody [clone: 336CT7.4.2]

Supplier: Abgent
AM1970A
89521-244EA 646.72 USD
89521-244
Anti-DICER1 Mouse Monoclonal Antibody [clone: 336CT7.4.2]
Antibodies
Western Blot: 1:500~2000,

Type: Primary
Antigen: DICER1
Clonality: Monoclonal
Clone: 336CT7.4.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR