Anti-SOX9 Mouse Monoclonal Antibody [clone: 334CT39.1.4]

Supplier: Abgent
AM1964A
89521-238EA 646.72 USD
89521-238
Anti-SOX9 Mouse Monoclonal Antibody [clone: 334CT39.1.4]
Antibodies
Western Blot: 1:1000~16000,

Type: Primary
Antigen: SOX9
Clonality: Monoclonal
Clone: 334CT39.1.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR