Anti-NCAM1 Mouse Monoclonal Antibody [clone: 321CT11.4.4]

Supplier: Abgent
AM1960A
89521-234EA 615.93 USD
89521-234
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 321CT11.4.4]
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:500~16000,

Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: 321CT11.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR