Anti-PGR Mouse Monoclonal Antibody [clone: 312CT8.2.1.6]

Supplier: Abgent
AM1958A
89521-232EA 646.72 USD
89521-232
Anti-PGR Mouse Monoclonal Antibody [clone: 312CT8.2.1.6]
Antibodies
Western Blot: 1:100~3200,

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 312CT8.2.1.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR