Anti-HSD3B1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP14585A
89521-162EA 646.4 USD
89521-162
Anti-HSD3B1 Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: 1:1000

Type: Primary
Antigen: HSD3B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR