Anti-BMI1 Mouse Monoclonal Antibody [clone: 353CT22.4.4]

Supplier: Abgent
AM1980B
89520-674EA 646.72 USD
89520-674
Anti-BMI1 Mouse Monoclonal Antibody [clone: 353CT22.4.4]
Antibodies
Western Blot: 1:200~1600,

Type: Primary
Antigen: BMI1
Clonality: Monoclonal
Clone: 353CT22.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR