Anti-COPS8 Rabbit Polyclonal Antibody

Supplier: Abgent
AP14987A
89520-644EA 646.4 USD
89520-644
Anti-COPS8 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: COPS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR