Anti-SNTB2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP16247C
89520-092EA 646.4 USD
89520-092
Anti-SNTB2 Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: 1:1000

Type: Primary
Antigen: SNTB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR