Anti-BMPR1A Rabbit Polyclonal Antibody

Supplier: Abgent
AP16478C
89519-722EA 646.4 USD
89519-722
Anti-BMPR1A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: BMPR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR