Anti-Troponin T type 3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP17019B
89518-998EA 646.4 USD
89518-998
Anti-Troponin T type 3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: Troponin T type 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR