Anti-ZWINT Mouse Monoclonal Antibody [clone: 422CT4.6.4]

Supplier: Abgent
AM2010B
89518-748EA 646.72 USD
89518-748
Anti-ZWINT Mouse Monoclonal Antibody [clone: 422CT4.6.4]
Antibodies
Western Blot: 1:100~500,

Type: Primary
Antigen: ZWINT
Clonality: Monoclonal
Clone: 422CT4.6.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR