Anti-LGR5 Mouse Monoclonal Antibody [clone: 349CT16.1.3.1]

Supplier: Abgent
AM1992B
89518-732EA 646.72 USD
89518-732
Anti-LGR5 Mouse Monoclonal Antibody [clone: 349CT16.1.3.1]
Antibodies
Western Blot: 1?2000,

Type: Primary
Antigen: LGR5
Clonality: Monoclonal
Clone: 349CT16.1.3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR