Anti-TLE1 Mouse Monoclonal Antibody [clone: 381CT18.5.1]

Supplier: Abgent
AM1982B
89518-724EA 646.72 USD
89518-724
Anti-TLE1 Mouse Monoclonal Antibody [clone: 381CT18.5.1]
Antibodies
Western Blot: 1:100~250,

Type: Primary
Antigen: TLE1
Clonality: Monoclonal
Clone: 381CT18.5.1
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR