Anti-CD7 Mouse Monoclonal Antibody [clone: 366CT28.2.3]

Supplier: Abgent
AM1974B
89518-714EA 646.72 USD
89518-714
Anti-CD7 Mouse Monoclonal Antibody [clone: 366CT28.2.3]
Antibodies
Western Blot: 1:100~250,

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 366CT28.2.3
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR