Anti-CLEC4D Rabbit Polyclonal Antibody

Supplier: Abgent
AP17517C
89518-240EA 646.4 USD
89518-240
Anti-CLEC4D Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CLEC4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR