Anti-TBR1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP17795A
89517-538EA 647.32 USD
89517-538
Anti-TBR1 Rabbit Polyclonal Antibody
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000

Type: Primary
Antigen: TBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR