Anti-CYORF15B Rabbit Polyclonal Antibody

Supplier: Abgent
AP17965C
89517-462EA 646.4 USD
89517-462
Anti-CYORF15B Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CYORF15B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR