Anti-CGB2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP17908A
89517-358EA 646.22 USD
89517-358
Anti-CGB2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CGB2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR