Anti-N6AMT1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP18103A
89517-282EA 646.4 USD
89517-282
Anti-N6AMT1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: N6AMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR