Anti-ALB Mouse Monoclonal Antibody [clone: 5CT2.1.4]

Supplier: Abgent
AM2069B
89516-756EA 646.72 USD
89516-756
Anti-ALB Mouse Monoclonal Antibody [clone: 5CT2.1.4]
Antibodies
Western Blot: 1:100~500,

Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: 5CT2.1.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR