Anti-CDC42EP1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP19010B
89515-536EA 646.4 USD
89515-536
Anti-CDC42EP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CDC42EP1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR