Anti-ST6GALNAC5 Rabbit Polyclonal Antibody

Supplier: Abgent
AP19473B
89514-798EA 646.4 USD
89514-798
Anti-ST6GALNAC5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: ST6GALNAC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR