Anti-HIST2H2BE Rabbit Polyclonal Antibody

Supplier: Abgent
AP19977A
89514-326EA 646.4 USD
89514-326
Anti-HIST2H2BE Rabbit Polyclonal Antibody
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000

Type: Primary
Antigen: HIST2H2BE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR