Anti-ARHGEF16 Rabbit Polyclonal Antibody

Supplier: Abgent
AP19783A
89514-094EA 646.4 USD
89514-094
Anti-ARHGEF16 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: ARHGEF16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR