Anti-PRPF39 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20140B
89513-872EA 646.4 USD
89513-872
Anti-PRPF39 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: PRPF39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR