Anti-PSMC1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20440B
89513-560EA 646.4 USD
89513-560
Anti-PSMC1 Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: 1:1000, Immunhistochemsitry: 1:25, Immunofluorescence: 1:25,

Type: Primary
Antigen: PSMC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR