Anti-ZRSR2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20436B
89513-548EA 646.72 USD
89513-548
Anti-ZRSR2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:25,

Type: Primary
Antigen: ZRSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR