Anti-H2AFX Mouse Monoclonal Antibody [clone: 980CT1.5.5]

Supplier: Abgent
AM2202B
89513-516EA 642.9 USD
89513-516
Anti-H2AFX Mouse Monoclonal Antibody [clone: 980CT1.5.5]
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:25,

Type: Primary
Antigen: H2AFX
Clonality: Monoclonal
Clone: 980CT1.5.5
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR