Anti-UCHL5 Mouse Monoclonal Antibody [clone: 854CT5.5.6]

Supplier: Abgent
AM2200B
89513-508EA 642.9 USD
89513-508
Anti-UCHL5 Mouse Monoclonal Antibody [clone: 854CT5.5.6]
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: UCHL5
Clonality: Monoclonal
Clone: 854CT5.5.6
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR