Anti-HOXC13A Rabbit Polyclonal Antibody

Supplier: Abgent
AZB10014A
89513-496EA 646.72 USD
89513-496
Anti-HOXC13A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: HOXC13A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR