Anti-ECE2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP20460C
89513-454EA 646.72 USD
89513-454
Anti-ECE2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:25,

Type: Primary
Antigen: ECE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR