Anti-TSC2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP3833A
89513-190EA 646.72 USD
89513-190
Anti-TSC2 Rabbit Polyclonal Antibody
Antibodies
Dot Blot: 1:500

Type: Primary
Antigen: TSC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR