Anti-LTBP1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7780A
89513-076EA 646.4 USD
89513-076
Anti-LTBP1 Rabbit Polyclonal Antibody
Antibodies
Immunhistochemsitry: 1:10~50

Type: Primary
Antigen: LTBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR