Anti-CDK5R1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7743B
89513-060EA 646.4 USD
89513-060
Anti-CDK5R1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50

Type: Primary
Antigen: CDK5R1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR