Anti-FOXD1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP14388A
89512-672EA 646.4 USD
89512-672
Anti-FOXD1 Rabbit Polyclonal Antibody
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:2000, Immunhistochemsitry: 1:10~50, Immunofluorescence: 1:10~50,

Type: Primary
Antigen: FOXD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR