Anti-CELA3A Rabbit Polyclonal Antibody

Supplier: Abgent
AP14493C
89512-554EA 646.4 USD
89512-554
Anti-CELA3A Rabbit Polyclonal Antibody
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000, Immunhistochemsitry: 1:10~50,

Type: Primary
Antigen: CELA3A
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR