Anti-TMPRSS13 Rabbit Polyclonal Antibody

Supplier: Abgent
AP14675B
89512-474EA 646.4 USD
89512-474
Anti-TMPRSS13 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50,

Type: Primary
Antigen: TMPRSS13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR