Anti-CTF1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP14905B
89512-442EA 646.4 USD
89512-442
Anti-CTF1 Rabbit Polyclonal Antibody
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000, Immunhistochemsitry: 1:10~50, Flow Cytometry: 1:10~50,

Type: Primary
Antigen: CTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR